Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Protein Kinase C-epsilon Translocation Inhibitor Peptide, Negative Control

Supplier: Thermo Scientific
J66521-EXD J66521-MCR
10814-670EA 75.07 USD InStock
10814-670 10814-668
Protein Kinase C-epsilon Translocation Inhibitor Peptide, Negative Control
Proteins and Peptides
Molecular Formula: C37H65N9O13
Formula Weight: 843.98
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White to off-white
Solubility: Soluble in water
MDL No.: MFCD07366708

A scrambled peptide with an identical amino acid composition to that of Protein Kinase C epsilon translocation Inhibitor Peptide. A cell impermeable negative control for PKC translocation inhibition.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty