Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pig Cecropin A

Supplier: Thermo Scientific
J66546-MCR J66546-EXD
10814-692EA 279.97 USD InStock
10814-692 10814-694
Pig Cecropin A
Proteins and Peptides
CAS No.: 80451-04-3
Molecular Formula: C184H313N53O46
Formula Weight: 4003.77
Storage Temperature: -30°C to -10°C
Physical Form: Lyophilized powder
Appearance: White to off-white
MDL No.: MFCD00130752

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty