Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

B-9430

Supplier: Thermo Scientific Chemicals

J66729-MCR J66729-EXD
10814-808EA 189.97 USD InStock
10814-808 10814-810
B-9430
B-9430


Formula: C₆₄H₉₅N₁₉O₁₃
MW: 1338.584 g/mol
Storage Temperature: Freezer
MDL Number: MFCD08705408
CAS Number: 180981-09-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty