Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Gentiobiose 98%

Supplier: Thermo Scientific Chemicals
Synonyms: β-D-Gentiobioseβ-Gentiobiose

J66750-MC J66750-ME J66750-03
10814-828EA 285.73 USD InStock
10814-828 10814-832 10814-830
Gentiobiose 98%
Gentiobiose


Formula: C₁₂H₂₂O₁₁
MW: 342.30 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00198056
CAS Number: 554-91-6
EINECS: 209-074-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty