Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Human [Glu1] Fibrinopeptide B

Supplier: Thermo Scientific
J66755-MCR
10814-834EA 130.65 USD InStock
10814-834
Human [Glu1] Fibrinopeptide B
Proteins and Peptides
CAS No.: 103213-49-6
Molecular Formula: C66H95N19O26
Formula Weight: 1570.57
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
Solubility: Soluble in water at 1mg/ml
MDL No.: MFCD00076440

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty