Would you like to log in again?
Plant Isolate DNA Extraction Kit, Thermo Scientific
Supplier: Thermo Scientific
J67858-AE
10816-208EA 307.41 USD InStock
10816-208
Plant Isolate DNA Extraction Kit, Thermo Scientific
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Plant isolate kit is CTAB and chloroform based method to isolate total DNA (including genomic, mitochondrial, and chloroplast DNA) from a variety of plant species (including algae and cyanobacteria). The extracted DNA is suitable for routine PCR screening, real-time PCR, southern blotting, mapping, and RFLP.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty