Would you like to log in again?
Air Track Adapter
Supplier: VERNIER SOFTWARE & TECH SE
ATA-DFS
470219-230EA 68.99 USD InStock
470219-230
Air Track Adapter
Educational Materials Physics Educational Materials Mechanics Learning Activities
Allows the Dual-Range Force Sensor to be mounted on the end of an air track for collision studies. Compatible with most air tracks distributed or made by PASCO scientific, Central Scientific, or Daedalon Corporation. Includes two magnetic bumpers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty