Would you like to log in again?
Quartz Cuvettes
Supplier: VERNIER SOFTWARE & TECH SE
CUV-QUARTZ
470219-264PK 298.99 USD InStock
470219-264
Quartz Cuvettes
Cuvettes Spectrophotometer Cuvettes
For use with UV-VIS Spectrophotometers
  • Standard rectangluar cuvettes
  • 170–2700nm spectral range
  • Two polished sides
The Quartz Cuvettes are a pack of two Spectrosil® Quartz cuvettes for use with UV-VIS spectrophotometers. They are the same pack of cuvettes that ship with the Vernier UV-VIS Spectrophotometer.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty