Would you like to log in again?
Microstirrer
Supplier: VERNIER SOFTWARE & TECH SE
MSTIR
470219-306EA 8.99 USD InStock
470219-306
Microstirrer
Stirrers Magnetic Stirrers
The Microstirrer takes the place of the stirring bar that is typically used with a laboratory magnetic stirrer. The new Vernier Microstirrer easily attaches to the end of a pH Sensor, Conductivity Probe, Dissolved Oxygen Probe, or Ion-Selective Electrode. When the magnetic stirrer plate (required, but not included) is turned on, its magnetic field turns the paddle wheel of the Microstirrer, resulting in enhanced stirring near the electrode tip. The Microstirrer works with any magnetic stirrer.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty