Would you like to log in again?
Standard-to-Mini USB Adapter
Supplier: VERNIER SOFTWARE & TECH SE
USB-MINI
470219-338EA 8.99 USD InStock
470219-338
Standard-to-Mini USB Adapter
Cables
This adapter allows a Go!Temp or Go!Link* to connect to the USB port of a TI-Nspire handheld or TI-84 graphing calculator. The adapter has a USB standard-A receptacle which connects to the Go!Temp or Go!Link and a USB Mini-A USB plug that connects to the calculator. The adapter can also be used with the USB cable that comes with a Go!Motion.
Connector 1: USB Standard Type A Receptacle
Connector 2: USB Mini Type A Plug
Most consumer electronic devices use a Mini-USB Type B plug, and will NOT WORK with this adapter. This adapter is only tested for use with TI-Nspire handhelds and TI graphing calculators.
* Due to calculator power limitations, only Go!Link revision 2 can be used with a TI-Nspire handheld or TI-84 graphing calculator. Go! Link revision 2 units are labeled “Vernier Go!Link”. Go!Link revision 1 units are labeled “Go!Link”.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty