Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Vernier® Circuit Board 2

Supplier: Vernier
VCB2
470219-368EA 144.99 USD InStock
470219-368
Vernier® Circuit Board 2
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
The Vernier Circuit Board 2 is a basic electricity lab on a board. The turret terminals for every component make it easy to connect basic series and parallel circuits, examine the behavior of different components, and investigate RLC circuits. The Vernier Circuit Board 2 includes 10 bulbs, a resettable fuse, three powering options, 10 alligator clip leads, terminal clips to add components, and pre-installed components.

Installed Components:
R1 and R2–10 Ω resistors rated at 5 W
R3 and R4–51 Ω resistors rated at 2 W
R5 and R6–68 Ω resistors rated at 2 W
R7–22 kΩ resistor rated at 1 W
R8–47 kΩ resistor rated at 1 W
R9–100 kΩ resistor rated at 1 W
R11–4.7 kΩ resistor rated at 1 W
C1–1 µF non-polar capacitor
C2–10 µF non-polar capacitor
3 E-10 lamp sockets
LED and 220 Ω current limiting resistor
L1–5 mH inductor, with powdered iron core in a clip
1 kΩ linear potentiometer
85 dB buzzer
SW2–single-pole, double-throw (SPDT) switch
SW3–single-pole, single-throw (SPST) momentary switch

Specifications:
Size: 10 in. x 6.5 in. (25.5 cm x 16.5 cm)
Mass: ~0.44 kg
Optional power sources:
up to 4 AA batteries, not included
LabQuest Power Supply, not included (or other 5 VDC adapter with center-positive ⅛ inch connector)
DC power supply such as Extech Digital DC power supply
Vernier Power Amplifier
Maximum voltage: 10 V
Resettable fuse opens at 0.8 A, but 0.3 A or less is recommended
All parts ROHS compliant

What’s Included:
One circuit board
Ten insulated jumper wires fitted with mini-alligator clips
Five #48 lamps rated 0.06 A at 2.0 V (cylindrical body)
Five #50 lamps rated 0.220 A at 7.5 V (round body)

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty