Would you like to log in again?
Barraquer Cilia Forceps, OR Grade, Sklar
Supplier: Sklar
66-1742
10194-412EA 180.8 USD InStock
10194-412
Barraquer Cilia Forceps, OR Grade, Sklar
Forceps
Barraquer Cilia Forceps are ophthalmologic instruments used for grasping or manipulating the cilia or eyelashes
These are thumb forceps with a stop peg to prevent an overly harsh grasp.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty