Would you like to log in again?
VWR® Pass-Thru Laboratory Refrigerators with Natural Refrigerant
Supplier: VWR
VWR®
CP-EW3-HCPTP CP-EW1-HCPTP HCPTP-47 CP-IOPQ-HCPTP CP-TM360-HCPTP HCPTP-49 CP-WIFI-HCPTP
10819-682EA 1703.84 USD InStock
10819-682 10819-684 10819-674 10819-680 10819-686 10819-676 10819-678
VWR® Pass-Thru Laboratory Refrigerators with Natural Refrigerant
Refrigerators
The VWR® Pass-Thru Laboratory Refrigerators come standard with microprocessor temperature controllers allowing for the precise temperature management necessary for critical cold storage.
  • Environmentally friendly refrigerants
  • Superior temperature uniformity
  • Remote alarm system
  • Self-maintenance keeps evaporator coils from freezing
The all new temperature display and alarm module offers a comprehensive array of features, providing up-to-the-minute sample simulated temperature information as well as the added notification features of the door ajar and power failure alarms
Forced-draft air circulation keeps cold air moving throughout the cabinet to ensure superior temperature uniformity and recovery after door openings. The cycle-defrost design allows for the self-maintenance necessary in all refrigerators to keep evaporator coils from freezing; but without the product-damaging spikes in temperature stemming from auto-defrost cycles. Height does not include 10.16cm (4") for casters.
Environmentally friendly, natural hydrocarbon (HC) refrigerants (not containing hydrofluorocarbons) vastly reduce global-warming potential (GWP) while meeting new EPA/SNAP mandates and UL, ASHRAE compliance guidelines.
Remote alarm contacts enable end users to tie these units into existing alarm systems to notify appropriate personnel of alarm events as soon as possible.
Certifications: UL/C-UL listed.
Ordering information: Units are supplied with a two-year parts and labor warranty, plus an additional two-year compressor parts warranty.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
COMPLEMENTARY PRODUCTS
SERVICES
Services: Factory temperature mapping on equipment to include 15 probes in air and 3 probes in sample solution for 360 minutes, with temperature readings every 60 seconds. Testing is conducted at manufacturer site. Validation Template with Temperature Mapping documentation helps customers execute validation on site after purchase. Must be purchased at the same time as the equipment.
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty