Would you like to log in again?
Human Recombinant FGF4 (from E. coli)
Supplier: BioVision
AB285468-1MG AB285468-25UG AB285468-100UG
10005-756EA 12448.85 USD InStock
10005-756 10005-758 10005-754
Human Recombinant FGF4 (from E. coli)
Proteins and Peptides
An important growth factor for fibroblasts and endothelial cells.
Recombinant human FGF-4 has full biological activity when compared standards. The ED₅₀, as determined by its ability to induce the proliferation of mouse NR6R-3T3 fibroblasts, is typically 0.25-1.25 ng/ml.
Fibroblast Growth Factor 4 (FGF-4) is a growth factor predominantly expressed during embryonic development, playing a key role in limb development. In culture, FGF-4 has been shown to be an important growth factor for fibroblasts and endothelial cells. Human FGF-4 shares high homology and cross-reactivity with the mouse protein. Recombinant human FGF-4, produced in E.coli, is a non-glycosylated protein containing 177 amino acids and having a total molecular mass of 19 kDa.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty