Would you like to log in again?
Human Recombinant IL2 (from E. coli)
Supplier: BioVision
AB285502-50UG AB285502-1MG AB285502-10UG
10006-122EA 154.44 USD InStock
10006-122 10006-126 10006-124
Human Recombinant IL2 (from E. coli)
Proteins and Peptides
A regulator of lymphocyte activities
The ED₅₀ as determined by the dose-dependent stimulation of murine CTLL-2 cells is < 0.1 ng/ml, corresponding to a specific activity of > 1 x 10⁷ units/mg.
IL-2 is a potent immuno-modulator, which mediates a wide range of immune and inflammatory responses. Recombinant Human IL-2 is a 15.5 kDa protein containing 134 amino acid residues.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty