Would you like to log in again?
Choice® Cleanroom Laundered 100% Polyester Knit Wipe, Berkshire
Supplier: Berkshire
CH709.14
89089-178CS 1018.07 USD InStock
89089-178
Choice® Cleanroom Laundered 100% Polyester Knit Wipe, Berkshire
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Ultrasonically sealed edge for reduced fiber contamination.
  • Chemically compatible with IPA, acetone and other solvents
Lightweight, high absorbency design can reduce landfill waste impact when compared to more traditional heavier weight products.
Applications include chamber cleaning and CMP processing, stencil and print roll cleaning, cleaning of medical device products, applying and removing cleaning and disinfecting solutions. Steam autoclavable for aseptic environments.
Certifications: Complies with RoHS and REACH requirements.
Ordering information: 150 sheets per pack, 14 packs per case. (double packed). Laundered and packaged in ISO class 4 (class 10) cleanroom.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty