Would you like to log in again?
Human Recombinant TNF alpha (from E. coli)
Supplier: BioVision
AB52126-10 AB52126-50 AB52126-1
10003-006EA 141.13 USD InStock
10003-006 10003-008 10003-010
Human Recombinant TNF alpha (from E. coli)
Proteins and Peptides
A potent cytokine exerting cytotoxic effects on a wide range of cells
The ED₅₀ as determined by the cytolysis of murine L929 cells in the presence of actinomycin D is < 0.05 ng/ml, corresponding to a specific activity of > 2 x 10⁷ units/mg.
Tumor Necrosis Factor-α(TNF-α) is a potent lymphoid factor that exerts cytotoxic effects on a wide range of tumor cells and certain other target cells. Human TNF-α is a 17.4 kDa protein containing 157 amino acid residues.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty