Would you like to log in again?
Human Recombinant Caspase 5 (from E. coli)
Supplier: BioVision
AB52109-100
10004-956EA 459.05 USD InStock
10004-956
Human Recombinant Caspase 5 (from E. coli)
Enzymes
A cysteine protease involved in apoptosis 
Caspase-5 (also know as ICErelIII, TY) is a member of the caspase-family of cysteine proteases
The recombinant active human caspase-5 was expressed in E. coli. The active caspase-5 is routinely tested at BioVision for its ability to enzymatically cleave these two substrates Ac-WEHD-pNA (Cat. #1102-200) or Ac-WEHD-AFC (1101-200).
Similar to other caspases, caspase-5 exists in cells as an inactive proenzyme which is matured by proteolysis to yield large and small subunits. The active caspase-5 is a heterotetramer consisting of two large and two small subunits. To date the mechanism of caspase-5 activation and roles of caspase-5 in apoptosis are poorly understood.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty