Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Human Recombinant Caspase 7 (from E. coli)
Supplier: BioVision
AB52173-25 AB52173-100
10005-026EA 281.55 USD InStock
10005-026 10005-024
Human Recombinant Caspase 7 (from E. coli)
Enzymes
A cysteine protease involved in apoptosis 
Caspase-7 (also know as Mch3, ICE-LAP3, CMH-1) is a member of the caspase-family of cysteine proteases
The recombinant active human caspase-7 was expressed in E. coli. The active caspase-7 is routinely tested at BioVision for its ability to enzymatically cleave these two substrates Ac-DEVD-pNA (Cat. #1008-200) or Ac-DEVD-AFC (1007-200).
Similar to other caspases, caspase-7 also exists in cells as an inactive proenzyme. During apoptosis procaspase-7 is processed at aspartate residues by self-proteolysis and/or cleavage by another caspase. The processed active form of caspase-7 consists of large and small subunits which associate to form the active enzyme. Active caspase-7 has been shown involving in the proteolysis of PARP (poly ADP-ribose polymerase), an enzyme that is involved in DNA repair and genomic maintenance.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty