Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Human Recombinant Caspase 8 (from E. coli)
Supplier: BioVision
AB54117-25
10005-030EA 281.98 USD InStock
10005-030
Human Recombinant Caspase 8 (from E. coli)
Enzymes
A cysteine-aspartic acid protease involved in apoptosis
Caspase-8 (also know as FLICE, MASH, Mch5) is a member of the caspase-family of cysteine proteases. Similar to other caspases, caspase-8 also exists in cells as an inactive proenzyme. During apoptosis procaspase-8 is processed at aspartate residues by self-proteolysis and/or cleavage by another caspase. The processed active form of caspase-8 consists of large and small subunits which associate to form the active enzyme. Active caspase-8 has been shown to activate caspase-3 leading to degradation of a variety of cellular target proteins during apoptosis. The recombinant active human caspase-8 was expressed in E. coli. The active caspase-8 is routinely tested at BioVision for its ability to enzymatically cleave these two substrates Ac-IETD-pNA (Cat. #1063-200) or Ac-IETD-AFC (1062-200).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty