Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Human Recombinant Caspase 10/B (from E. coli)
Supplier: BioVision
AB52081-100
10005-036EA 452.49 USD InStock
10005-036
Human Recombinant Caspase 10/B (from E. coli)
Enzymes
A cysteine protease involved in apoptosis 
Caspase-10b (also known as FLICE 2) is a member of the caspase-family of cysteine proteases. Similar to other caspases, caspase-10 also exists in cells as an inactive proenzyme. During apoptosis procaspase-10 is processed at aspartate residues to form active caspase-10 which consists of two large (18 kDa) and two small (11 kDa) subunits in a heterotetramer form. The recombinant active human caspase-10/b was expressed in E. coli. The active caspase-10 is routinely tested at BioVision for its ability to enzymatically cleave these two substrates Ac-IETD-pNA (Cat. #1063-200) or Ac-IETD-AFC (1062-200).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty