Would you like to log in again?
Econo™ Troeltsch (Wilde) Forceps, Floor Grade, Sklar
Supplier: Sklar
Econo™
21-227
89023-084PK 123.08 USD InStock
89023-084
Econo™ Troeltsch (Wilde) Forceps, Floor Grade, Sklar
Forceps
Econo™ Troeltsch (Wilde) Forceps are thumb forceps used in ENT procedures.
They may be known as Troeltsch forceps, or Wilde-Troeltsch forceps. These distinctive forceps feature a 80° angle for packing and dressing in nasal and ear procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty