Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Econo™ Bozeman Forceps, Floor Grade, Sklar
Supplier: Sklar
Econo™
21-374
89023-184PK 233.8 USD InStock
89023-184
Econo™ Bozeman Forceps, Floor Grade, Sklar
Forceps
Econo™ Bozeman Forceps are a multi-functional instrument used in gynecological procedures
The shanks of this instrument are slightly curved, and provide visibility when used in the uterine cavity. The jaws are available either curved or straight. This product features an "S" curve (slightly curved shanks with jaws slightly curved opposite direction) with partial horizontal serrations.
Econo™ instruments are disposable.
They are used for tissue removal in dilation and curettage procedures, intrauterine device retrieval, and packing the uterine cavity.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty