Would you like to log in again?
Econo™ Rochester-Oschner Forceps, Floor Grade, Sklar
Supplier: Sklar
21-515 21-542 21-513 21-504 21-521 21-500 21-488
76192-594BX 170.36 USD InStock
76192-594 89023-228 76192-600 76192-598 76192-588 76192-596 76192-574
Econo™ Rochester-Oschner Forceps, Floor Grade, Sklar
Forceps
Econo™ Rochester-Oschner Forceps are a heavy instrument designed to aggressively grasp medium to heavy tissue or occlude heavy, dense vessels.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
The combination of full serrations plus teeth ensure a firm grip on the tissue or vessel being held. Rochester-Oschner Forceps are available either curved or straight and in multiple lengths. These forceps are sometimes referred to as simply Oschner Forceps, however, Rochester-Oschner Forceps are actually heavier than standard Oschner Forceps.
Econo™ instruments are disposable.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty