Would you like to log in again?
Sklarlite XD™ Adson Dressing Forceps, OR Grade, Sklar
Supplier: Sklar
23-2604
10194-222EA 111.27 USD InStock
10194-222
Sklarlite XD™ Adson Dressing Forceps, OR Grade, Sklar
Forceps
Sklarlite XD™ Adson Dressing Forceps are thumb forceps used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds
They have a wide thumb grasp for increased precision and control. Sklarlite XD™ instruments are noticeably lighter and delicate than standard instruments which allow the surgeon the highest degree of precision with the least amount of fatigue. These instruments have been uniquely manufactured to be one of the lightest, most delicate general instruments available today. This product is straight with serrated tips and a length of 4-3/4 inches
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty