Would you like to log in again?
Human Recombinant Cathepsin L (from E. coli)
Supplier: BioVision
AB81780-5
10005-112EA 327.38 USD InStock
10005-112
Human Recombinant Cathepsin L (from E. coli)
Proteins and Peptides
Lysosomal cysteine protease involved in protein degradation, arthritis, apoptosis, and cancer
> 10000 mU/mg (1 U = digestion of 1 µmole of Z-Phe-Arg-AFC substrate (K142-100) per minute at 37°C in 50 mM Sodium Acetate, 100 mM NaCl, pH 5.5).
Cathepsin L is a lysosomal protease that can be secreted from the cell and is implicated to play a role in protein degradation, arthritis, apoptosis, and cancer.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty