Would you like to log in again?
Mouse Recombinant Caspase 3 (from E. coli)
Supplier: BioVision
AB52071-100
10005-124EA 393.47 USD InStock
10005-124
Mouse Recombinant Caspase 3 (from E. coli)
Enzymes
A cysteine-aspartic acid protease involved in apoptosis
Caspase-3 (also know as CPP32, Yama and apopain) is a major member of the caspase-family of cysteine proteases. Caspase-3 exists in cells as an inactive 32 kDa proenzyme. During apoptosis procaspase-3 is processed at aspartate residues by self-proteolysis and/or cleavage by upstream caspases, such as caspase-6 (Mch2), caspase-8 (Flice) and grazyme B. The processed form of caspase-3 consists of large (17 kD) and small (11 kD) subunits which associate to form the active enzyme. The active caspase-3 has been shown involving in the proteolysis of several important molecules, such as poly (ADP-ribose) polymerase (PARP), the sterol regulatory element binding proteins (SREBPs), focal adhesion kinase (FAK), and others. The recombinant active human caspase-3 expressed in E. coli spontaneously undergoes autoprocessing to yield subunits characteristic of the native enzyme. The active caspase-3 preferentially cleaves caspase-3 substrates (e.g., DEVD-AFC or DEVD-pNA) and is routinely tested at BioVision for its ability to enzymatically cleave these two substrates Ac-DEVD-pNA (Cat. #1008-200) or Ac-DEVD-AFC (1007-200).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty