Would you like to log in again?
SatPax® 670-R Nonwoven Cleanroom Wiper, Berkshire
Supplier: Berkshire
SPX670R00112
89107-784CS 464.67 USD InStock
89107-784
SatPax® 670-R Nonwoven Cleanroom Wiper, Berkshire
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
SatPax® 670-R combines Durx® 670 non woven wipers composed of a hydroentangled nonwoven blend of 55% cellulose and 45% polyester with a high saturation level.
  • Low extractables and fiber and particle counts
  • High abrasion resistance
  • Reduces alcohol usage and preparation/handling costs
  • Increases cleaning efficiency
  • Smooth and durable with good wet strength
Designed for use in wet cleaning of critical surfaces, where control of flammable solvents and flammable solvent concentrations is required. Reduces VOC (Volatile Organic Compound) emissions.
ISO Class 5 and higher cleanroom environments and applications requiring USP 797 compliance. Re-sealable solvent resistant packaging. Final cleaning of surfaces or products prior to manufacturing or packaging. High saturation level for removing cleaning, and disinfecting residues in regulated environments.
Certifications: Complies with RoHS and REACH requirements.
Ordering information: 75 sheets per pack, 12 packs per case (double packed).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty