Would you like to log in again?
Barraquer Needle Holder, OR Grade, Sklar
Supplier: Sklar
65-6457 65-6456 65-6454 65-6455 65-6450 65-6445 65-6449 65-6452 65-6446
10194-614EA 779.41 USD InStock
10194-614 10194-602 10194-616 10194-604 10194-596 10194-610 10194-598 10194-612 10194-600
Barraquer Needle Holder, OR Grade, Sklar
Forceps
Sklar's® Barraquer Needle Holder is used for holding needles in microsurgical and ophthalmic procedues where primarily fine needles are used
It has a spring handle, commonly with a slightly raised and rounded part that has a smooth grip for improved control. It is available with or without a locking mechanism.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty