Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Barraquer Wire Speculum, OR Grade, Sklar
Supplier: Sklar
65-1756 65-1758 65-1744 65-1742
10194-630EA 108.93 USD InStock
10194-630 10194-640 10194-632 10194-642
Barraquer Wire Speculum, OR Grade, Sklar
Specula
Sklar's® Barraquer Wire Speculum is an ophthalmic instrument used in retracting the eye lid and holding it open during surgery.
  • Self-retaining retractor
This speculum is a lightweight, spring style, wireform instrument, therefore minimal pressure is placed on the eye.
Delivery information: Available with solid or fenestrated blades.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty