Would you like to log in again?
Castroviejo Needle Holder, OR Grade, Sklar
Supplier: Sklar
65-6350 65-6358 65-6354 65-6352 65-6357
10197-932EA 833.13 USD InStock
10197-932 10197-414 10194-066 10197-930 10197-416
Castroviejo Needle Holder, OR Grade, Sklar
Forceps
Sklar's Castroviejo Microsurgical Needle Holder is a double-spring instrument used for holding small, delicate needles in various microsurgical procedures
Censitrac Ready products are industry standard and constructed of stainless steel.
This device is curved with smooth jaws. It also features a locking mechanism.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty