Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Surgi-OR™ Pederson Vaginal Speculum, Mid Grade, Sklar
Supplier: Sklar
95-362 95-347 95-348
89023-590EA 57.14 USD InStock
89023-590 89023-602 89023-592
Surgi-OR™ Pederson Vaginal Speculum, Mid Grade, Sklar
Specula
Surgi-OR™ Pederson Vaginal Speculum is a gynecological instrument used for performing pelvic examinations, Pap Smears, and examining the cervix.
It is a double blade duckbill speculum inserted with the blades closed. The blades are then opened by an external lever and screw. The blades of the Pederson Speculum are more narrow than the blades of the Graves Speculum. The practitioner can best determine which type and size speculum is most appropriate for the patient.
Hand-crafted in Pakistan, Surgi-OR™ mid-grade instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty