Would you like to log in again?
Econo™ Sterile Strabismus Scissors, Sklar
Supplier: Sklar
96-2542 96-2543 96-2541 96-2540
89023-826CS 602.04 USD InStock
89023-826 89023-830 89023-832 89023-828
Econo™ Sterile Strabismus Scissors, Sklar
Scissors
Econo™ Sterile instruments are ready-to-use, sterile, disposable, floor grade instruments made in Pakistan
Econo™ Sterile instruments eliminate the need for cleaning and reprocessing, as they are both sterile and disposable. Econo™ Sterile instruments significantly reduce the likelihood of cross-contamination. They are available in bulk quantities.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty