Would you like to log in again?
LiteGrip™ Bishop-Harmon Forceps, OR Grade, Sklar
Supplier: Sklar
66-4134 66-4034
10197-502EA 345.31 USD InStock
10197-502 10194-400
LiteGrip™ Bishop-Harmon Forceps, OR Grade, Sklar
Forceps
Sklar LiteGrip™ Bishop Harmon forceps are thumb forceps commonly used in ophthalmic procedures
They have long, narrow jaws for handling fine tissues found around the eye. LiteGrip™ instruments feature a lightweight, fenestrated handle that is beneficial to use during lengthy procedures when fatigue may arise. The finger holes on the handle also allow grip adjustments as needed by the surgeon.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty