Would you like to log in again?
Sklar Splinter Removal Kit, OR Grade, Sklar
Supplier: Sklar
19-3360
10195-040EA 172.81 USD InStock
10195-040
Sklar Splinter Removal Kit, OR Grade, Sklar
Forceps
The Sklar Splinter Removal Kit is used for exposing and removing splinters or other small foreign objects such as ticks
This kit contains one Carmalt Splinter Forceps and one Vanguard Splinter Pick. The splinter pick is used to widen the area in which the object is lodged in order for the forceps to grasp and remove it. This kit includes a storage case. The Carmalt Splinter forceps are straight and 4½" long. The Vanguard Splinter Pick is 4 1/2" long with a 3 1/4" handle.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty