Would you like to log in again?
Human Recombinant IL21 (from E. coli)
Supplier: BioVision
AB285535-50UG AB285535-1MG AB285535-10UG
10006-338EA 8564.81 USD InStock
10006-338 10006-336 10006-340
Human Recombinant IL21 (from E. coli)
Proteins and Peptides
>90% Pure active Recombinant Human Interleukin-21 (IL-21)
The recombinant human IL-21 is fully biologically active when compared to standard. The ED₅₀ as determined by the dose-dependent stimulation of human peripheral blood mononuclear cells is <5 ng/ml.
Recombinant human IL-21 is a homodimeric, non-glycosylated polypeptide containing 133 amino acid residues and has a molecular weight of 15.5 kDa.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty