Would you like to log in again?
Human Recombinant TPO (from E. coli)
Supplier: BioVision
AB285559-10UG AB285559-1MG AB285559-50UG
10006-602EA 1262.1 USD InStock
10006-602 10006-600 10006-598
Human Recombinant TPO (from E. coli)
Proteins and Peptides
>98% Pure active Recombinant human TPO.
The ED₅₀ as determined by the dose-dependent stimulation of the proliferation of human MO7e cells was found <1 ng/ml, corresponding to a specific activity of >1 x 10⁶ IU/mg.
TPO (Thrombopoietin, also known as Mpl-ligand, MGDF) stimulates the proliferation and maturation of megakaryocytes and promotes increased circulating levels of platelets in vivo. TPO signals through the c-mpl receptor and acts as an important regulator of circulating platelets. Human and murine TPO exhibits cross-species reactivity. Recombinant human TPO is a fully biologically active 174 amino acid polypeptide, which contains the erythropoietin-like domain of the full length TPO protein.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty