Would you like to log in again?
Cat Recombinant SDF1 beta (from E. coli)
Supplier: BioVision
AB125767-10UG
10006-674EA 643.53 USD InStock
10006-674
Cat Recombinant SDF1 beta (from E. coli)
Proteins and Peptides
>97% Pure active Recombinant feline SDF-1beta (CXCL12).
The activity is determined by its ability to chemoattract human T cells at 10 - 75 ng/ml.
Stromal Derived Growth Factor-1beta (SDF-1beta), also called CXCL12, is one of two splice variants made by a wide variety of cells when stimulated by inflammatory cytokines such as, TNF, IL-1 or LPS. SDF-1β signals through the G protein-couple receptor, CXCR4, to recruit activated leukocytes. Human SDF-1β shares 100% identity to feline SDF-1β. Recombinant feline SDF-1beta is a non-glycosylated protein, comprised of 72 amino acids, with a molecular weight of 8.5 kDa.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty