Would you like to log in again?
VWR® THree-Channel Electronic Timer and Clock with Certificate of Calibration
Supplier: VWR
VWR®
61700-3211
82023-860CS 1375.39 USD InStock
82023-860
VWR® THree-Channel Electronic Timer and Clock with Certificate of Calibration
Timers Digital Timers
This 3-channel timer has a capacity of 19 hours, 59 minutes and 59 seconds or can be switched to a 12- or 24-hour clock.
  • Counts either up or down
  • Last time set memory
  • Multiple mounting options
Features a Certificate of Calibration and individual serial number, these electronic timers are tested and calibrated in H-B’s exclusive triple accredited/registered ISO/IEC 17025:2005, A2LA accredited laboratory (Cert #2448.01), against equipment whose calibration is traceable to NIST.
Certifications: CE marked, RoHS compliant.
Ordering information: Requires two LR44 batteries (included).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty