Would you like to log in again?
Human Recombinant Rage (from Cells)
Supplier: BioVision
AB138789-50UG AB138789-1MG AB138789-10UG
10006-272EA 10694.53 USD InStock
10006-272 10006-270 10006-274
Human Recombinant Rage (from Cells)
Proteins and Peptides
>95% Pure Recombinant Human RAGE.
Advanced glycosylation end product-specific receptor, also known as receptor for advanced glycosylation end products, AGER and RAGE, belongs to the immunoglobulin superfamily of cell surface molecules. It lies within the major histocompatibility complex (MHC) class III region on chromosome 6. Besides AGEs, AGER is also able to bind other ligands which is thought to result in pro-inflammatory gene activation. It is known that AGER serves as a mediator of both acute and chronic vascular inflammation in certain conditions such as atherosclerosis and in particular as a complication of diabetes. Furthermore, it plays an important role in regulating the production/expression of TNF-alpha, oxidative stress, and endothelial dysfunction in type 2 diabetes.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty