Would you like to log in again?
Annexin V-Pe Apoptosis Det Kit, BioVision
Supplier: BioVision
AB14155-100
10013-274EA 545.27 USD InStock
10013-274
Annexin V-Pe Apoptosis Det Kit, BioVision
Assays Protein Assays
Detects Apoptosis in Live Cells within 10 min.
  • Flow cytometry (Ex = 488 nm; Em = 578 nm) and fluorescence microscopy
  • Mammalian
  • Convenient sizes (25, 100, 400 assays)
  • Detect early/middle stages of apoptosis; differentiate apoptosis from necrosis
The Annexin V-PE Apoptosis Detection Kit is based on the observation that soon after initiating apoptosis, most cell types translocate the membrane phospholipid phosphatidylserine (PS) from the inner face of the plasma membrane to the cell surface. Once on the cell surface, PS can be easily detected by staining with a fluorescent conjugate of Annexin V, a protein that has a strong natural affinity for PS. The one-step staining procedure takes only 10 minutes. In addition, the assay can be directly performed on live cells, without the need for fixation.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty