Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cathepsin D Inhibitor Screening Kit, BioVision

Supplier: BioVision
AB126779-100TEST
10013-604EA 1225.74 USD InStock
10013-604
Cathepsin D Inhibitor Screening Kit, BioVision
Assays Protein Assays
Simple Assay, HTS

  • Fluorescence (Ex/Em = 328/460 nm)
  • 100 assays
  • Assay kit to detect putative Cathepsin D inhibitors

Apoptosis can be mediated by mechanisms other than the traditional caspase-mediated cleavage cascade. There is growing recognition that alternative proteolytic enzymes such as the lysosomal cathepsin proteases may initiate or propagate proapoptotic signals. Cathepsins are lysosomal enzymes that are also used as sensitive markers in various toxicological investigations. The Cathepsin D Activity Assay kit is a fluorescence-based assay that utilizes the preferred cathepsin D substrate sequence GKPILFFRLK(Dnp)-D-R-NH2) labeled with MCA. Cathepsin D will cleave the synthetic substrate to release the quenched fluorescent group MCA, which can then easily be measured using a fluorometer or fluorescence plate reader at Ex/Em = 328/460 nm. The relative efficacy of test inhibitors are compared to the positive control inhibitor, Pepstatin A (IC50 < 0.1 nM). The Cathepsin D assay is simple, straightforward, and can be adapted to 96-well plate assays and is suitable for high throughput screening (HTS).

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty