Would you like to log in again?
Caspglow Act Caspa-3 Stain Kit, BioVision
Supplier: BioVision
AB65613-100
10013-638EA 603.14 USD InStock
10013-638
Caspglow Act Caspa-3 Stain Kit, BioVision
Assays Protein Assays
Detects activated Caspase-3 by FACS, FL or plate reader
  • Fluorescence microscopy, Flow cytometry and fluorescence plate reader (Ex. = 485 nm and Em. = 535 nm)
  • Mammalian
  • Convenient sizes (25 and 100 assays)
  • Sensitive detection of activated caspase-3 in living cells
Activation of caspases plays a central role in apoptosis. The CaspGLOW™ Fluorescein Active Caspase-3 Staining Kit provides a convenient means for sensitive detection of activated caspase-3 in living cells. The assay utilizes the caspase-3 inhibitor, DEVD-FMK, conjugated to FITC (FITC-DEVD-FMK) as a marker. FITC-DEVD-FMK is cell permeable, nontoxic, and irreversibly binds to activated caspase-3 in apoptotic cells.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty