Would you like to log in again?
Glutathione Fluorometric Assay Kit, BioVision
Supplier: BioVision
AB65322-100
10013-660EA 944.25 USD InStock
10013-660
Glutathione Fluorometric Assay Kit, BioVision
Assays Protein Assays
Simple assay to detect total glutathione
  • Fluorescence (Ex/Em 380/461 nm)
  • Mammalian
  • The Glutathione Detection Kit provides a simple in vitro assay for detecting the GSH changes in apoptosis and other pathological processes
Glutathione is the principal intracellular low-molecular-weight thiol that plays a critical role in the cellular defense against oxidative and nitrosative stress in mammalian cells. Diminished glutathione levels have been observed in the early stages of apoptosis. BioVision’s ApoGSH™ Glutathione Detection Kit provides a simple in vitro assay for detection of total glutathione changes in apoptosis and other samples. The assay utilizes monochlorobimane (MCB), a dye that appears to form an adduct exclusively with glutathione. The unbound MCB is almost nonfluorescent, whereas the dye fluoresces blue (Ex./Em. = 380nm/461nm) when bound to glutathione of reduced or oxidized form. The reaction is catalyzed by glutathione S-transferase. Thus, the amount of total glutathione can be easily detected using a fluorometer or a 96-well fluorometric plate reader.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty