Would you like to log in again?
Cytosol/Particulate Separation Kit, BioVision
Supplier: BioVision
AB65398-50
10013-666EA 1072.98 USD InStock
10013-666
Cytosol/Particulate Separation Kit, BioVision
Assays Protein Assays
Rapidly separates cytosol from particulate fractions
  • Mammalian
  • The Cytosol/Particulate Separation Kit provides an easy and convenient system for a rapid separation of cytosol and particulate fractions. Assaying cellular distributions of small moleculars or proteins, and other signal transduction pathways
The location and translocation of proteins, signaling molecules, and other small molecules inside cells regulate cell growth, differentiation and many other cellular functions. Separating cytosol and particulate fractions is an important step in studying subcellular localization of cellular components. However, traditional methods usually take hours to separate cytosol from particulates, so that some cellular components especially small molecules and metabolites are getting diffused or redistributed during the separation procedure. The new Cytosol/Particulate Rapid Separation Kit physically separate cytosol from particulate compartments rapidly through an oil layer and thus the two fractions would not contact or diffuse to each other. Using the method, contaminations can be avoided even for small molecules. Subcellular localization and analyses of factors interested can be performed accurately.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty