Would you like to log in again?
Immunoprecipitation Ip Kit, BioVision
Supplier: BioVision
AB206996-25TEST
10013-670EA 708.29 USD InStock
10013-670
Immunoprecipitation Ip Kit, BioVision
Assays Protein Assays
Only Kit on the market for both IP/Co-IP of proteins/complexes as well as their functional study and analysis
  • Immunoprecipitation (IP) and Co-IP
  • Functional study of Immunoprecipitated proteins/complexes
  • SDS-PAGE or western blot analysis of Immunoprecipitated proteins/complexes
Immunoprecipitation (IP) is widely used in research and development, by which a protein can be selectively purified from samples. BioVision’s immunoprecipitation kit provides optimized buffers for preparing cell/tissue extracts, antigen binding and washing steps. The protein A/G Sepharose beads provided in the kit has higher binding capacity with broader antibody isotype binding than traditional protein A or protein G resins. The kit can be used in variety of immunoprecipitation or Co-IP studies.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty