Would you like to log in again?
Bioluminescence Cytotoxicity Assay Kit, BioVision
Supplier: BioVision
AB228557-500TEST
10013-676EA 374.8 USD InStock
10013-676
Bioluminescence Cytotoxicity Assay Kit, BioVision
Assays Protein Assays
Cytotoxicity assay based on adenylate kinase (AK), HTS
  • Luminescence
  • Mammalian
Cell death or cytotoxicity is classically evaluated by the quantification of plasma membrane damage. Adenylate kinase (AK) is a ubiquitous protein present in all eukaryotic and prokaryotic cells and rapidly release into culture medium upon damage of the plasma membrane. The Bioluminescence Cytotoxicity Assay Kit is based on the measurement of AK in a simple one-step procedure involving two chemical reactions. The first reaction is the conversion of ADP to ATP by adenylate kinase that was released from the damaged cells. The second reaction utilizes luciferase to catalyze the formation of light from ATP and luciferin, and the light can be measured using a luminometer or beta counter. The assay is highly sensitive and can be fully automatic for high throughput.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty