Would you like to log in again?
Mouse Recombinant IL20 (from E. coli)
Supplier: BioVision
AB51977-10
10006-764EA 281.98 USD InStock
10006-764
Mouse Recombinant IL20 (from E. coli)
Proteins and Peptides
>95% Pure Recombinant murine IL-20.
The biological activity was determined by measuring the dose-dependent activation of STAT3 in human epidermal keratinocytes (HEK2a cells). Significant STAT3 activation is observed with ≥100 ng/ml of recombinant mouse IL-20.
Mouse IL-20 is fully biologically active when compared to standard. The biological activity was determined by measuring the dose-dependent activation of STAT3 in human epidermal keratinocytes (HEK2a cells). Significant STAT3 activation is observed with >100 ng/ml of recombinant mouse IL-20
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty