Would you like to log in again?
Cathepsin D Activity Assay Kit, BioVision
Supplier: BioVision
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
AB65302-100
10013-596EA 911.15 USD InStock
10013-596
Cathepsin D Activity Assay Kit, BioVision
Assays Protein Assays
  • Fluorescence (Ex/Em 328/460)
  • Mammalian
  • 100 assays
  • Detects alternative proteolytic enzymes such as the lysosomal cathepsin proteases that can initiate or propagate proapoptotic signals
Apoptosis can be mediated by mechanisms other than the traditional caspase-mediated cleavage cascade. There is growing recognition that alternative proteolytic enzymes such as the lysosomal cathepsin proteases may initiate or propagate proapoptotic signals. Cathepsins are lysosomal enzymes that are also used as sensitive markers in various toxicological investigations. The Cathepsin-D Activity Assay kit is a fluorescence-based assay that utilizes the preferred cathepsin-D substrate sequence GKPILFFRLK(Dnp)-D-R-NH2) labeled with MCA. Cell lysates or other samples that contain cathepsin-D will cleave the synthetic substrate to release fluorescence, which can then easily be quantified using a fluorometer or fluorescence plate reader at Ex/Em = 328/460 nm.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty