Would you like to log in again?
Human Recombinant BMP7 (from E. coli)
Supplier: BioVision
AB285600-10UG AB285600-50UG AB285600-1MG
10006-804EA 359.53 USD InStock
10006-804 10006-808 10006-806
Human Recombinant BMP7 (from E. coli)
Proteins and Peptides
>98% Pure active Recombinant human BMP-7.
BioVision’s recombinant human BMP-7 is biologically active. The ED₅₀ as determined by dose-dependent induction of alkaline phosphatase activity with C2C12 cells is in the range of 2-12 µg/ml.
BMPs (bone morphogenetic proteins) belong to the TGF-β superfamily of structurally related signaling proteins. As implied by their name, BMPs promote and regulate bone development, growth, remodeling and repair, in both prenatal development and postnatal growth of eye, heart, kidney, skin, and other tissues. Recombinant human BMP-7 produced from E. coli is a 15.7 kDa monomeric, non-glycosylated, polypeptide chain containing 139 amino acid residues.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty